Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rhotekin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA593182
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA593182 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA593182 Supplier Invitrogen™ Supplier No. PA593182
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PLCMTQPTAS GTLRVQQAGE MQNWAQVHGV LKGTNLFCYR QPEDADTGEE PLLTIAVNKE TRVRAGELDQ ALGRPFTLSI SNQYGDDEVT HTLQTESREA LQSWMEALWQ LFFDMSQWKQ CCDEIMKIET PAPRKPPQAL AKQGSLYHEM AIEPLDDIAA VTDILTQREG ARLETPPPWL AMFTDQPALP NPCSPASVAP APDWTHPLPW GRPRTFSLDA VPPDHSPRAR SVAPLPPQRS PRTRGLCSKG QPRTWLQSPV; Positive Samples: A-431, HepG2, MCF7.

This gene encodes a scaffold protein that interacts with GTP-bound Rho proteins. Binding of this protein inhibits the GTPase activity of Rho proteins. This protein may interfere with the conversion of active, GTP-bound Rho to the inactive GDP-bound form by RhoGAP. Rho proteins regulate many important cellular processes, including cytokinesis, transcription, smooth muscle contraction, cell growth and transformation. Dysregulation of the Rho signal transduction pathway has been implicated in many forms of cancer. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rhotekin
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.14 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RTKN
Gene Accession No. Q6V7V2, Q8C6B2, Q9BST9
Gene Alias hypothetical protein LOC539691; RGD:727874}; rhotekin; RTKN; rtkn {ECO:0000312; RTKN1
Gene Symbols RTKN
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 254-513 of human RTKN (NP_0010150561).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20166, 297383, 6242
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.