Learn More
Eurofins Discoverx Recombinant Human IL-23, 10 µg. (92-1266)
Supplier:  Eurofins Discoverx 921266
Encompass_Preferred
Interleukin 23 (IL-23) is a cytokine protein and mediator of the immune response. IL-23 is composed of IL-23A and the p40 subunit of IL-12 (IL-12B). Il-23 is an agonist of the receptor formed by IL-12RB1 and IL-23R responsible for activation of STAT4 and production of IFNG, and acts preferentially on CDR(+) T cells. IL-23 activates the JAK-STAT pathway, promoting production of pro-inflammatory cytokines and has been implicated in autoimmune inflammatory diseases and tumorigenesis. Provided as 10 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9785
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.