Learn More
Eurofins Discoverx Recombinant Human Relaxin-2, 5 µg. (92-1203)
Supplier: Eurofins Discoverx 921203
Relaxin-2 is a protein hormone structurally similar to insulin, and is the only relaxin that circulates in the blood. Binds specifically to the receptors RXFP1 (LGR7) and RXFP2 (LGR8). Plays an important role in female reproduction, and may have a role in enhancing sperm motility, regulating blood pressure, controlling heart rate, and releasing oxytocin and vasopressin. Provided as 5 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter, PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.