Learn More
R Biopharm Inc RIDASCREEN FAST Milk
Supplier:  R Biopharm Inc R4652
Encompass
RIDASCREEN FAST Milk is a sandwich enzyme immunoassay to quantify milk proteins (casein and beta-lactoglobulin) in food containing whey, milk or milk powder such as sausages, ice cream, chocolate, bakery goods, cake and bread mix, soups, sauces, dressings and beverages (juice, wine, beer). The antibodies specifically detect caseins as well as beta-lactoglobulin of cow’s milk and of sheep’s, goat’s and buffalo’s milk. The assay results are quantitative if the sample contains milk or milk powder.
Catalog No. NC1926975
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.