Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIAM Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501728787 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8787 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8787 Supplier Proteintech Group Inc Supplier No. 135001AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RIAM appears to function in the signal transduction from Ras activation to actin cytoskeletal remodeling. Suppresses insulin-induced promoter activities through AP1 and SRE. Mediates Rap1-induced adhesion.

Specifications

Antigen RIAM
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.59 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene APBB1IP
Gene Accession No. Q7Z5R6
Gene Alias APBB1 interacting protein 1, APBB1IP, INAG1, PREL 1, PREL1, Proline rich EVH1 ligand 1, Proline rich protein 73, RARP 1, RARP1, RIAM
Gene Symbols APBB1IP
Host Species Rabbit
Immunogen RIAM Fusion Protein Ag4327
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54518, 54519
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.