Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ribavirin Monoclonal Antibody (1D8)
GREENER_CHOICE

Catalog No. PIMA549659
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549659 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549659 Supplier Invitrogen™ Supplier No. MA549659
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

Ribavirin is an antiviral medication that is used to treat a variety of viral infections, including hepatitis C and respiratory syncytial virus (RSV). It works by inhibiting viral replication and preventing the virus from spreading to other cells in the body. Ribavirin is typically used in combination with other antiviral drugs to increase its effectiveness.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ribavirin
Applications ELISA, Immunohistochemistry, Lateral Flow
Classification Monoclonal
Clone 1D8
Concentration 8 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Host Species Mouse
Immunogen Conjugate of RBV hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.