Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ribavirin, USP, 98.9-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18606075 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-075 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-606-075 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. R1052500GM

missing translation for 'validMainframeMsg'

R1052, 36791-04-5, C8H12N4O5

Spectrum™ Chemical Ribavirin, USP is an antiviral drug used primarily to treat hepatitis C and viral hemorrhagic fevers. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 36791-04-5
Molecular Formula C8H12N4O5
Molecular Weight (g/mol) 244.21
InChI Key IWUCXVSUMQZMFG-AFCXAGJDSA-N
IUPAC Name 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1H-1,2,4-triazole-3-carboxamide
SMILES NC(=O)C1=NN(C=N1)[C@@H]1O[C@H](CO)[C@@H](O)[C@H]1O

Specifications

CAS Max % 1
Ignition Residue 0.0025
Loss on Drying 0.005
Packaging Amber Glass Bottle
Quantity 500 g
Specific Rotation -33.5° to -37.0°
Percent Purity 98.9 to 101.5%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.