CAS RN 36791-04-5
Ribavirin Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Analog of the imidazole nucleotide intermediates in purine nucleotide biosynthesis.
Ribavirin Impurity Standard, MilliporeSigma™ Supelco™
Analog of the imidazole nucleotide intermediates in purine nucleotide biosynthesis.
Ribavirin, 98%
CAS: 36791-04-5 Molecular Formula: C8H12N4O5 Molecular Weight (g/mol): 244.2 InChI Key: IWUCXVSUMQZMFG-AFCXAGJDSA-N Synonym: ribavirin,tribavirin,virazole,rebetol,ribavirine,copegus,vilona,ribamide,ribamidil,ribasphere PubChem CID: 37542 ChEBI: CHEBI:63580 IUPAC Name: 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,4-triazole-3-carboxamide SMILES: C1=NC(=NN1C2C(C(C(O2)CO)O)O)C(=O)N
Ribavirin, MP Biomedicals™
CAS: 36791-04-5 Molecular Formula: C8H12N4O5 Molecular Weight (g/mol): 244.207 InChI Key: IWUCXVSUMQZMFG-AFCXAGJDSA-N Synonym: ribavirin,tribavirin,virazole,rebetol,ribavirine,copegus,vilona,ribamide,ribamidil,ribasphere PubChem CID: 37542 ChEBI: CHEBI:63580 IUPAC Name: 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,4-triazole-3-carboxamide SMILES: C1=NC(=NN1C2C(C(C(O2)CO)O)O)C(=O)N
Ribavirin 98.0+%, TCI America™
CAS: 36791-04-5 Molecular Formula: C8H12N4O5 Molecular Weight (g/mol): 244.207 MDL Number: MFCD00058564 InChI Key: IWUCXVSUMQZMFG-AFCXAGJDSA-N Synonym: ribavirin,tribavirin,virazole,rebetol,ribavirine,copegus,vilona,ribamide,ribamidil,ribasphere PubChem CID: 37542 ChEBI: CHEBI:63580 IUPAC Name: 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,4-triazole-3-carboxamide SMILES: C1=NC(=NN1C2C(C(C(O2)CO)O)O)C(=O)N
R&D Systems™ Ribavirin
CAS: 36791-04-5 Molecular Formula: C8H12N4O5
Ribavirin, USP, 98.9-101.5%, Spectrum™ Chemical
CAS: 36791-04-5 Molecular Formula: C8H12N4O5 Molecular Weight (g/mol): 244.21 InChI Key: IWUCXVSUMQZMFG-AFCXAGJDSA-N IUPAC Name: 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1H-1,2,4-triazole-3-carboxamide SMILES: NC(=O)C1=NN(C=N1)[C@@H]1O[C@H](CO)[C@@H](O)[C@H]1O