Learn More
Marketlab, Inc. Ribbed Tube Caps for 12 mm Tubes
Description
- Designed with multiple ribs to provide a leak-proof seal
- Constructed of flexible polyethylene
- Grooved top eases grip for easy on/off
Specifications
Specifications
| Product Type | Test Tube Plug Cap |
| Autoclavable | Not Autoclavable |
| Material | Polyethylene |
| Color | Blue |
| Sterility | Non-sterile |
| Disposable | Single-use |