Learn More
Market Lab Ribbed Tube Caps For 12mm Tubes, Blue
Supplier:  Market Lab 1433BL
Encompass_Preferred
Designed with multiple ribs to provide a leak-proof seal. Constructed of flexible polyethylene. Grooved top eases grip for easy on/off.
Catalog No. NC1690541
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.