Learn More
Essendant Fargo Multi Color Thermal Resin Printer Ribbon, Black/Cyan/Magenta/Yellow
Supplier:  Essendant SRX45010
Encompass_Preferred
The Fargo 45010 YMCKOK Color Ribbon is a premium quality ribbon for Fargo DTC1000 and DTC1250 plastic card printers It is a full-color dye with an overlay panel for extra protection Plus this ribbon also comes with two sets of black resin dye that enable dual-sided printing and are essential for producing readable barcodes Device Types- Printer Color(s)- Black Cyan Magenta Yellow OEM/Compatible- OEM Ribbon Type- Multicolor Thermal Resin
Catalog No. 50-000-27886
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.