Learn More
Essendant Magicard 300 Dual YMCKO/2 Printer Ribbon, Black/Cyan/Magenta/Yellow
Supplier:  Essendant SRXMC300YMCKO2
Encompass_Preferred
Magicard consumables are especially designed for Magicard printers to ensure crisp high-quality images and text Tested and approved by Magicard Rand D genuine Magicard consumables offer the best performance and an extended lifespan for your printer Magicard 300 YMCKO/2 color ribbon has yellow (Y) magenta (M) and cyan (C) panels for creating full-color images on one side of an ID card The black resin (K) panel is for printing black text images and barcodes on the same side of the card The overlay (O) panel will apply a clear layer over the printed side of the card to protect the images and text from fading Device Types- Printer Color(s)- Black Cyan Magenta Yellow OEM/Compatible- OEM Ribbon Type- YMCKO Color
Catalog No. 50-000-27394
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.