Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Ribonuclease A, bovine pancreas, chromatographically purified

Catalog No. AAJ62232EX3
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62232EX3 200 mg
AAJ6223203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62232EX3 Supplier Thermo Scientific Chemicals Supplier No. J62232EX3
Add to Cart

missing translation for 'validMainframeMsg'

Used to remove RNA from DNA plasmid preparations and protein samples. Ribonuclease A is used for RNase protection assays, to remove unspecifically bound RNA, analysis of RNA sequences, to hydrolyze RNA contained in protein samples, and the purification of DNA.

Ribonuclease A is used to remove RNA from DNA plasmid preparations and protein samples. Ribonuclease A is used for RNase protection assays, to remove unspecifically bound RNA, analysis of RNA sequences, to hydrolyze RNA contained in protein samples, and the purification of DNA. Ribonuclease A from bovine pancreas has been used in a study to assess hybridase activity of human ribonuclease-1. Ribonuclease A from bovine pancreas has also been used in a study to investigate particle-based and monolithic columns for cation exchange protein displacement chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Sensitivity Moisture sensitive
Quantity 200 mg
Form Lyophilized

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.