Learn More
Sigma Aldrich Fine Chemicals Biosciences PIPES 99 titration1KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences P67571KG
Encompass
PIPES is a member of the ethanesulfonic acid buffer series first introduced by Good et al. developed to meet certain criteria midrange pKa maximum water solubility and minimum solubility in all other solvents minimal salt effects minimal change in pKa with temperature chemically and enzymatically stable minimal absorption in visible or UV spectral range and easily synthesized. Since its pKa at 37 C is near physiological pH PIPES has applications in cell culture work.
Catalog No. 50-291-4642
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.