Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ribonucleic acid from Baker's yeast

Catalog No. AAJ6121503
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6121503 1 g
AAJ61215MC 100 mg
AAJ61215ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6121503 Supplier Thermo Scientific Chemicals Supplier No. J6121503
Add to Cart

CAS: 63231-63-0

Ribonucleic acid (RNA) from baker?s yeast S. cerevisiae may be used as a substrate for studying ribonuclease activities of enzymes such as RNAase(s). It is also suitable for use as a carrier in nucleic acid purification and precipitation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ribonucleic acid (RNA) from baker′s yeast S. cerevisiae may be used as a substrate for studying ribonuclease activities of enzymes such as RNAase(s). It is also suitable for use as a carrier in nucleic acid purification and precipitation.

Solubility
It is soluble in water.

Notes
Prepared by a modification of Crestfield, A.M., et al., J. Biol. Chem., 216, 185 (1955). Suitable for use as a ribonuclease substrate. Not suitable for use as a carrier in nucleic acid purification and precipitation
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63231-63-0
MDL Number MFCD00132196
IUPAC Name Ribonucleic acid

Specifications

Color White to Brown
Quantity 1 g
Solubility Information It is soluble in water.
Physical Form Powder
Chemical Name or Material Ribonucleic acid from Baker's yeast
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.