Learn More
AdipoGen Ribosomal Protein S3 h pAb
SDP
Supplier:  AdipoGen AG25A0077C100
Encompass_Preferred
Polyclonal Antibody. Recognizes human ribosomal protein S3. Detects a band of ~32kDa by Western blot. Source Rabbit. Applications ELISA, ICC, IHC, IP, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. RpS3 is a component of the 40S ribosomal subunit and is an an essential but previously unkown subunit of NF-kappaB involved in the regulation of key genes in rapid cellular activation responses. RpS3 interacts with nm23-H1. The expression of rpS3 reduces the secretion of MMP-9 and the invasive metastatic potential in HT1080 cells. The phosphorylated ERK is reduced by the expression of rpS3.
Catalog No. 50-256-3238
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.