Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIC3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501735756 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5756 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5756 Supplier Proteintech Group Inc Supplier No. 277761AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Promotes functional expression of homomeric alpha-7 and alpha-8 nicotinic acetylcholine receptors at the cell surface. May also promote functional expression of homomeric serotoninergic 5- HT3 receptors, and of heteromeric acetylcholine receptors alpha- 3/beta-2, alpha-3/beta-4, alpha-4/beta-2 and alpha-4/beta-4.

Specifications

Antigen RIC3
Applications Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene RIC3
Gene Accession No. Q7Z5B4, Q8BPM6
Gene Alias AYST720, PRO1385, Protein RIC 3, RIC3
Gene Symbols Ric3
Host Species Rabbit
Immunogen RIC3 Fusion Protein Ag26740
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 320360, 79608
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.