Learn More
Epredia™ Richard-Allan Scientific™ Gram Stain (Tissue) Kit, Crystal Violet Solution
Catalog No. 23005260 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Crystal Violet
Decolorizing Solution
Gram Stain (Tissue) Kit
Safranin O Stain Solution
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
23-005-260 Safranin O Stain Solution
22-110-647 Gram Stain (Tissue) Kit
23-005-259 Crystal Violet
23-005-261 Decolorizing Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 23-005-260 Supplier Epredia™ Supplier No. 88103
Add to Cart
Add to Cart
This item is not returnable. View return policy
Identify gram-positive and gram-negative bacteria in tissue sections with Epredia™ Richard-Allan Scientific™ Gram Stain (Tissue)
  • Gram-positive organisms stain blue to blue-black
  • Gram-negative organisms stain red
  • Tartrazine provides a yellow background stain
Specifications
Quantity 500mL
Product Type Safranin O Stain Solution
WARNING: Reproductive Harm - www.P65Warnings.ca.gov