Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ High-Quality Dehydrants

Catalog No. 9515 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 gal.
1 gal., 4/case
5 gal.
Product Type:
Acetone
Ethyl Alcohol
Isopropyl Alcohol
Methyl Alcohol
Reagent Alcohol
Concentration:
100%
70%
92%
99%
IPA:5.5%, EtOH:90%, MeOH:4.5%
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type Concentration
95-15 5 gal. Isopropyl Alcohol 99%
22-050-131 1 gal., 4/case Acetone 100%
22-050-132 1 gal., 4/case Ethyl Alcohol 92%
9570 1 gal., 4/case Isopropyl Alcohol 70%
22-050-134 1 gal., 4/case Isopropyl Alcohol 99%
22-050-135 1 gal., 4/case Methyl Alcohol 99%
9990520 1 gal. Reagent Alcohol IPA:5.5%, EtOH:90%, MeOH:4.5%
91-11 1 gal., 4/case Reagent Alcohol IPA:5.5%, EtOH:90%, MeOH:4.5%
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 95-15 Supplier Epredia™ Supplier No. 9515
Add to Cart
Add to Cart

Reduce spills with easy-to-pour Epredia™ High-Quality Dehydrants.

  • Formulated to rigid specifications,providing laboratories with high-quality products
  • Packaged in high-density polyethylene, which is safer and lighter than glass

Specifications

Boiling Point 81° to 83°C
Color Colorless
Compatibility Incompatible: Strong oxidizing agents, Acids, Halogens, Acid anhydrides
Content And Storage Keep containers tightly closed in a dry, cool and well-ventilated place. Keep away from heatand sources of ignition. Flammables area. Keep containers tightly closed in a cool, wellventilatedplace. Keep in properly labeled containers
Concentration 99%
Melting Point -89.5°C
Product Type Isopropyl Alcohol
Quantity 5 gal.
Form Liquid
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.