Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Mark-It™ Tissue Marking Dyes

Catalog No. 22050460 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Tissue Marking Dye
Tissue Marking Dye Kit
Color:
Black
Blue
Blue, Green, Orange, Black, Red, Yellow
Green
Orange
Red
Yellow
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Color
22-050-460 Tissue Marking Dye Yellow
22-050-456 Tissue Marking Dye Black
22-110-752 Tissue Marking Dye Blue
22-050-458 Tissue Marking Dye Green
22-050-660 Tissue Marking Dye Orange
22-050-459 Tissue Marking Dye Red
22-050-455 Tissue Marking Dye Kit Blue, Green, Orange, Black, Red, Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 22-050-460 Supplier Epredia™ Supplier No. 5000Y
Add to Cart
Add to Cart
This item is not returnable. View return policy

For a variety of tissue identification purposes use Epredia™ Mark-It™ Tissue Marking Dyes.

Features:
  • Permanent dyes that maintain their original brilliance during routine tissue processing cycles
  • Can be used on fresh or formalin-fixed tissues

Uses include:

  • Marking surgical margins
  • Identifying serial segments of large tissues to ensure proper orientation and embedding
  • Maintaining proper tissue orientation for Mohs surgery specimens
  • Identification of multiple pieces of tissues processed in the same cassette

Ordering Information:

  • Available in black, blue, green, orange, red and yellow in 2 oz. bottles

Specifications

Color Yellow
Product Type Tissue Marking Dye
Quantity 2 oz./ ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.