Learn More
Epredia™ Richard-Allan Scientific™ Modified Grocott's Methenamine Silver (Chromic Acid) Kit
Shop All Epredia ProductsDescription
- Utilizes methenamine borax capsules to make a working silver solution
- Distinguishes fungi, basement membranes, and opportunistic organisms in tissue sections
- Chromic acid solution enhances detection of Pneumocystis carinii, histoplasmosis, and fungus organisms; eliminates non-specific background staining
- Entities and reticular fibers stain black against a green background
Specifications
Specifications
| For Use With (Application) | Histological and Cytological Staining |
| Includes | 12 Methenamine Borax Capsules, 125 mL Fast Green Stain Solution, 125 mL Sodium Thiosulfate Solution, 500 mL Sodium Metabisulfite Solution, 125 mL Gold Chloride Solution, 500 mL 5% Chromic Acid Solution, 30 mL 5% Silver Nitrate Solution |
| Quantity | 1 Kit |
| Product Type | Stain Kit |