Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RICTOR Monoclonal Antibody (454313)
GREENER_CHOICE

Catalog No. MA561458
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA561458 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA561458 Supplier Invitrogen™ Supplier No. MA561458
  • $447.00
    $335.65 / Each of 1

    Save $111.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

Reconstitute at 0.5 mg/mL in sterile PBS.

RICTOR and MTOR (FRAP1) are components of a protein complex that integrates nutrient- and growth factor-derived signals to regulate cell growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RICTOR
Applications Western Blot
Classification Monoclonal
Clone 454313
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with trehalose and no preservative
Gene Rictor
Gene Accession No. Q6R327
Gene Alias 4921505C17Rik; 6030405M08Rik; AVO3; AVO3 homolog; AW492497; D530039E11Rik; DKFZp686B11164; Fyb; FYN binding protein; hAVO3; Kiaa1999; mAVO3; MGC39830; PIA; pianissimo; protein pianissimo; Rapamycin-insensitive companion of mTOR; rictor; RPTOR independent companion of MTOR complex 2; RPTOR independent companion of MTOR, complex 2; TORC2-specific protein AVO3
Gene Symbols Rictor
Host Species Rat
Immunogen E. coli-derived recombinant human Rictor, Met1-Leu250.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 253260
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.