Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ Ridgeline™ Bump Cap

Catalog No. 19155427 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Color:
Black
Blue
Brown
Gray
Green
Orange
Red
White
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-155-427 Brown
19-155-424 White
19-155-425 Black
19-155-426 Gray
19-155-428 Red
19-155-429 Yellow
19-155-430 Green
19-155-431 Orange
19-155-432 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19-155-427 Supplier Pyramex™ Supplier No. HP40015
Add to Cart
Add to Cart

Ridgeline™ Bump Cap provides stylish head protection from minor bumps and scalp lacerations.

  • Stylish Ridgeline bump caps are available for applications where hard hats are not required, but some protection from minor bumps and scalp lacerations are needed
  • Ridgeline bump caps are made of polyethylene material and are not designed to meet the safety standards for industrial head protection
  • This cap is not designed to meet the ANSI Z89.1-2009 class C, G, & E standards for industrial head protection
  • Custom imprinting available

Specifications

Color Brown
Dimensions (L x W x H) 27.5 x 19.5 x 14 cm
For Use With (Application) Where hardhats are not required, but some protection from minor bumps and scalp lacerations are needed
Material Shell: HDPE, Sweat Band: Polyester fabric, Polyurethane foam
Product Type Bump Cap
Quantity 1 Ea.
Size 6.5 to 8
Suspension Type 4-point glide lock suspension
Weight (Metric) 215 g
Ventilated Not Vented
Head Style Short Brim (Cap Style)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.