Learn More
Description
- ABS material – strong, yet ultra-light for seemingly weightless protection
- Low profile design – offers a low center of gravity for better balance
- Rear padded suspension – provides additional level of comfort on the wearers’ neck
- 4-position harness points allows you to move the harness forward, backward, up or down – allowing for the most optimal position for
ultimate comfort - Replaceable sweat band – padded fabric and polyurethane foam for all day wear
- Vented pressure pad keeps the straps in place providing an additional level of comfort
- Easy to grasp knob adjusts the suspension from 6 1/2 to 8
Specifications
Specifications
| Certifications/Compliance | ANSI Z89.1 standards, Type 1 --- Class C, G, and E |
| Color | Hi-vis Lime |
| For Use With (Application) | Construction |
| Material | Shell: ABS, Suspension: HDPE and high impact Nylon, Sweat band: Polyester Fabric and Polyurethane Foam |
| Product Type | 4-Point Ratchet |
| Size | 6.5 to 8 |
| Weight (Metric) | 358 g |
| Ventilated | Not Vented |
| Head Style | Short Brim (Cap Style) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.