Learn More
Description
- Shell constructed from ABS materials
- Ratchet suspension is easy to adjust and allows the wearer to modify the
fit while wearing hard hat - Replaceable suspensions and headbands available
- 4-point suspension may be converted to 6-point suspension with
optional strap (HP6PTSUS) - Custom imprinting available on hard hats
Specifications
Specifications
| Certifications/Compliance | ANSI Z89.1 standards, Type 1 --- Class C, G, and E,CSA Z94.1-05 |
| Color | Black |
| For Use With (Application) | Construction |
| Material | Shell: ABS, Suspension: HDPE and high impact Nylon, Sweat band: Polyester Fabric and Polyurethane Foam |
| Product Type | 4-Point Ratchet |
| Size | 6.5 to 8 |
| Weight (Metric) | 378 g |
| Ventilated | Not Vented |
| Head Style | Full Brim |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.