Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ Ridgeline™ Full Brim Hard Hat

Catalog No. 19155472 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Color:
Black
Blue
Green
Hi-vis Lime
Hi-vis Orange
Hi-vis Pink
Matte Black Graphite
Matte Camo
Matte Copper
Red
Slate Gray
White
White Graphite
Yellow
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-155-472 Yellow
19-155-464 White
19-155-465 Black
19-155-466 Slate Gray
19-155-467 White Graphite
19-155-468 Matte Black Graphite
19-155-469 Matte Copper
19-155-470 Matte Camo
19-155-471 Red
19-155-473 Hi-vis Lime
19-155-474 Green
19-155-475 Hi-vis Orange
19-155-476 Blue
19-155-477 Hi-vis Pink
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. 19-155-472 Supplier Pyramex™ Supplier No. HP54130
Add to Cart
Add to Cart

Ridgeline™ Full Brim Hard Hat is strong, yet ultra-light for seemingly weightless protection.

  • Shell constructed from ABS materials
  • Ratchet suspension is easy to adjust and allows the wearer to modify the
    fit while wearing hard hat
  • Replaceable suspensions and headbands available
  • 4-point suspension may be converted to 6-point suspension with
    optional strap (HP6PTSUS)
  • Custom imprinting available on hard hats

Specifications

Certifications/Compliance ANSI Z89.1 standards, Type 1 --- Class C, G, and E,CSA Z94.1-05
Color Yellow
For Use With (Application) Construction
Material Shell: ABS, Suspension: HDPE and high impact Nylon, Sweat band: Polyester Fabric and Polyurethane Foam
Product Type 4-Point Ratchet
Size 6.5 to 8
Weight (Metric) 378 g
Ventilated Not Vented
Head Style Full Brim
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.