Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex Safety Products Ridgeline™ Full Brim Hard Hat, 4-Point Ratchet Suspension

Catalog No. 19555102 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Color:
Gray
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-555-102 Gray
19-555-104 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-555-102 Supplier Pyramex Safety Products Supplier No. HP54112
Add to Cart
Add to Cart

Ridgeline™ Full Brim Hard Hat with 4-Point Ratchet Suspension is easy to adjust and allows the wearer to modify the fit while wearing hard hat.

  • Shell is constructed from ABS materials
  • Soft brow pad is replaceable
  • Replaceable suspension and headband also available

Specifications

Certifications/Compliance ANSI Z89.1
Color Gray
For Use With (Application) Construction
Material ABS
Product Type 4-point Ratchet Suspension
Size 6.5 to 8
Ventilated Not Vented
Head Style Full Brim
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.