Learn More
Pyramex™ Ridgeline™ Hydro Dipped Capstyle Hard Hat
Catalog No. 19155438 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Color:
Matte Black Graphite
Matte Copper
White Graphite
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-155-438 White Graphite
19-155-440 Matte Black Graphite
19-155-442 Matte Copper
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-155-438 Supplier Pyramex™ Supplier No. HP44116S
Only null left
Add to Cart
Add to Cart
Strong, yet ultra-light for seemingly weightless protection
  • ABS material – strong, yet ultra-light for seemingly weightless protection
  • Low profile design – offers a low center of gravity for better balance
  • Rear padded suspension – provides additional level of comfort on the
    wearers’ neck
  • 4-position harness points allows you to move the harness forward,
    backward, up or down – allowing for the most optimal position for
    ultimate comfort
  • Replaceable sweat band – padded fabric and polyurethane foam for
    all day wear
  • Vented pressure pad keeps the straps in place providing an additional
    level of comfort
  • Easy to grasp knob adjusts the suspension from 6 ½ to 8
  • Specifications
    Certifications/Compliance ANSI Z89.1 standards, Type 1 --- Class C, G, and E
    Color White Graphite
    Finish Glossy
    Material Shell: ABS, Suspension: HDPE and high impact Nylon, Sweat band: Polyester Fabric and Polyurethane Foam
    Product Type 4-Point Ratchet
    Weight (Metric) 358 g