Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex Safety Products Ridgeline™ Vented Full Brim Hard Hat

Catalog No. 19555106 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Color:
Blue
Gray
White
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-555-106 Yellow
19-555-101 White
19-555-103 Gray
19-555-107 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-555-106 Supplier Pyramex Safety Products Supplier No. HP54130V
Add to Cart
Add to Cart

Ridgeline™ Vented Full Brim Hard Hat is strong, yet ultra-light for seemingly weightless protection.

  • ABS material
  • Ratchet suspension is easy to adjust and allows the wearer to modify the fit while wearing hard hat
  • Soft brow pad is replaceable
  • Replaceable suspensions and headbands also available
  • Custom imprinting available on hard hats
  • 4-point suspensions can be converted to a 6-point suspension with the HP6PTSUS
  • Meets ANSI Z89.1-2014, Cert. Mod. - CAN/CSA Z94.1-15, Type 1, Class E, G, and C

Specifications

Certifications/Compliance Ansi Z89.1
Color Yellow
For Use With (Application) Construction
Product Type 4-point Ratchet Suspension
Size 6.5 to 8
Suspension Type 4-point ratchet suspension
Ventilated Vented
Head Style Full Brim
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.