Learn More
Medchemexpress LLC Rifabutin (standard) | 72559-06-9 | MFCD00866816 | 99.0% | 10 MG
Supplier: Medchemexpress LLC HY1702510MG
Rifabutin is a semisynthetic ansamycin antibiotic supplied as a research and analytical standard. It inhibits DNA-dependent RNA polymerase and is commonly used in vitro to study antimycobacterial activity or as a reference compound in analytical assays. The material is provided as a solid with high purity appropriate for laboratory use.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.