Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rifampicin, Fisher BioReagents™

Catalog No. BP26795 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Amber Glass
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP26795 5 g Amber Glass
BP26791 1 g Amber Glass
BP267925 25 g Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP26795

missing translation for 'validMainframeMsg'

Rifampicin

  • An antibiotic used to inhibit synthesis of proteins during recombinant protein expression in bacteria
  • Rif inhibits bacterial DNA-dependent RNA polymerase 
  • Rif can also be used as a selective agent for some strains
  • Mammalian RNA polymerase is not affected
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13292-46-1
Molecular Formula C43H58N4O12
Molecular Weight (g/mol) 822.953
InChI Key FZYOVNIOYYPUPY-HRJPTAQKSA-N
Synonym Rif
PubChem CID 131839595
SMILES CC1C=CC=C(C(=O)NC2=C(C3=C(C(=C4C(=C3C(=O)C2=CNN5CCN(CC5)C)C(=O)C(O4)(OC=CC(C(C(C(C(C(C1O)C)O)C)OC(=O)C)C)OC)C)C)O)O)C

Specifications

Type Rifampicin
Physical Form Solid
Melting Point 183°C
Loss on Drying < 1.0%
Packaging Amber Glass
Color Orange
Quantity 5 g
Assay Percent Range 97%
Merck Index 15, 8341
Formula Weight 822.96
Chemical Name or Material Rifampicin
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Substances known to cause developmental toxicity in humans. Harmful if swallowed. May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required.

NFPA

Health:2
Flammability:0
Instability:0
Recommended Storage 4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.