accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHNONHNNCH3OOOOHCH3OHCH3OOH3CCH3OH3CH3CH3CHOHOH3C(E/Z)(E/Z)(E/Z)(E/Z)

CAS RN 13292-46-1

IUPAC Name: 2,15,17,29-tetrahydroxy-11-methoxy-3,7,12,14,16,18,22-heptamethyl-26-{[(4-methylpiperazin-1-yl)amino]methylidene}-6,23,27-trioxo-8,30-dioxa-24-azatetracyclo[23.3.1.1⁴,⁷.0⁵,²⁸]triaconta-1,3,5(28),9,19,21,25(29)-heptaen-13-yl acetate
Synonyms: rifampicin
Molecular Weight (g/mol): 822.95
Molecular Formula: C43H58N4O12
InChi Key: FZYOVNIOYYPUPY-UHFFFAOYNA-N
SMILES: COC1C=COC2(C)OC3=C(C)C(O)=C4C(O)=C(NC(=O)C(C)=CC=CC(C)C(O)C(C)C(O)C(C)C(OC(C)=O)C1C)C(=CNN1CCN(C)CC1)C(=O)C4=C3C2=O
Quantity 
  • (5)
  • (1)
  • (1)
  • (1)
  • (5)
  • (1)
  • (5)
  • (1)
Physical Form 
  • (1)
  • (1)
  • (9)
  • (5)
Type 
  • (1)
  • (14)
Color 
  • (3)
  • (5)
  • (1)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (5)
  • (1)
  • (7)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (5)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (9)
  • (5)
  • (1)
  • (14)
  • (3)
  • (5)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)