Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rifampicin, Molecular Biology Grade, G-Biosciences
SCI_ED

Catalog No. S139836
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139836 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139836 Supplier G-Biosciences Supplier No. RC867
Add to Cart
Add to Cart

Rifampicin is active against gram-positive bacteria but less active against gram-negative bacteria.

It interferes with the synthesis of nucleic acids by inhibiting DNA dependent RNA polymerase. Resistance to Rifampicin can develop rapidly. The degree of resistance depends on the site of mutation in the RNA polymerase.

Chemical Identifiers

CAS 13292-46-1

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Powder
Sterility Non-sterile
Color White
Quantity 100 g
Percent Purity 98%
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.