Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rifampin, 100.3%, MP Biomedicals™

Catalog No. ICN19549025 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19549025 25 g
ICN19549001 1 g
ICN19549005 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19549025 Supplier MP Biomedicals, Inc Supplier No. 0219549025
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13292-46-1
Molecular Formula C43H58N4O12
Molecular Weight (g/mol) 822.953
InChI Key FZYOVNIOYYPUPY-HRJPTAQKSA-N
Synonym Rifampicin
PubChem CID 131839595
SMILES CC1C=CC=C(C(=O)NC2=C(C3=C(C(=C4C(=C3C(=O)C2=CNN5CCN(CC5)C)C(=O)C(O4)(OC=CC(C(C(C(C(C(C1O)C)O)C)OC(=O)C)C)OC)C)C)O)O)C

Specifications

Type Rifampicin
Physical Form Powder
Color Orange/Red
Quantity 25 g
Assay Percent Range 1.003
Formula Weight 823
Chemical Name or Material Rifampin
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.