Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Rifampin, 95%

Catalog No. AC455620050
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455620050 5 g
AC455620010 1 g
AC455620250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC455620050 Supplier Thermo Scientific Chemicals Supplier No. 455620050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

Rifampin (rifampicin), CAS # 13292-46-1, is a synthetic rifamycin antibiotic that works by inhibiting RNA and DNA synthesis and exhibits activity against the tuberculosis microbe.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Rifampin is a rifamycin class antibiotic synthesized to resemble the compound isolated from Amycolatopsis rifamycinica

Applications

• Rifampin can be used as a bacteriostatic that prevents RNA and DNA synthesis, particularly in mycobacteria

• It may also act as an antineoplastic, a neuroprotective, and inhibitor of angiogenesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13292-46-1
Molecular Formula C43H58N4O12
Molecular Weight (g/mol) 822.953
InChI Key FZYOVNIOYYPUPY-HRJPTAQKSA-N
PubChem CID 131839595
SMILES CC1C=CC=C(C(=O)NC2=C(C3=C(C(=C4C(=C3C(=O)C2=CNN5CCN(CC5)C)C(=O)C(O4)(OC=CC(C(C(C(C(C(C1O)C)O)C)OC(=O)C)C)OC)C)C)O)O)C

Specifications

Type Rifampicin
Melting Point 183.0°C
Infrared Spectrum Authentic
Loss on Drying 2% max. (100 mg, 60°C, 4 h) (vacuum)
Quantity 5 g
Solubility Information Solubility in water: slightly soluble.
Formula Weight 822.94
Percent Purity 95%
Chemical Name or Material Rifampin
Hazard Category Warning
Hazard Statement GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Do not eat, drink or smoke when using this product. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.