Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rifampin, MP Biomedicals™

Catalog No. MP021954901 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021954901 100 mg
MP021954902 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP021954901 Supplier MP Biomedicals, Inc Supplier No. 02195490.1
Add to Cart
Add to Cart

Inhibits DNA-dependent bacterial RNA polymerase. Rifampin, MP Biomedicals is a bactericidal antibiotic drug of the rifamycin group that does not affect mammalian RNA polymerase. It is used to treat Tuberculosis and Tuberculosis-related mycobacterial infec

Mode of Action: Inhibits initiation of RNA synthesis by binding to β-subunit of RNA polymerase. It inhibits the assembly of DNA and protein into mature virus particles.

  • Treats Tuberculosis and Tuberculosis-related mycobacterial infections
  • Widely used as an antipruritic agent in the autoimmune cholestatic liver disease, primary biliary cirrhosis (PBC)
  • It has been shown to cause hepatitis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13292-46-1
Molecular Formula C43H58N4O12
Molecular Weight (g/mol) 822.953
InChI Key FZYOVNIOYYPUPY-HRJPTAQKSA-N
Synonym Rifamycin AMP, Rifampicin, 3-(4-Methylpiperazinyliminomethyl)rifamycin SV, Rifampicin
PubChem CID 131839595
SMILES CC1C=CC=C(C(=O)NC2=C(C3=C(C(=C4C(=C3C(=O)C2=CNN5CCN(CC5)C)C(=O)C(O4)(OC=CC(C(C(C(C(C(C1O)C)O)C)OC(=O)C)C)OC)C)C)O)O)C

Specifications

Physical Form Powder
Absorbance λ max (methanol): 237–239, 334–338, 475–479nm
Melting Point 183°C to 188°C (decomposition)(literature)
Extinction Coefficient EmM (max absorbance, phosphate buffer, pH 7.38): 33.20 (237nm); 32.10 (255nm); 27.00 (334nm);15.40 (475nm) (Lit.)
Quantity 100 mg
Optical Rotation [α]D25°=+10.6° (c=0.5% in CDCl3)(Lit.)
Solubility Information Soluble in chloroform (349mg/mL, 25°C), DMSO (∽100mg/mL), ethyl acetate (108mg/mL, 25°C), methanol (16mg/mL, 25°C) tetrahydrofuran.
Formula Weight 822.96
Recommended Storage +4°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.