Learn More
AdipoGen Rifamycin AF-K56035
SDP
Supplier:  AdipoGen AGCN20323M050
Encompass_Preferred
Chemical. CAS 39976-84-6. Formula C44H52N4O14. MW 860.9. Semisynthetic. Ansamycin antibiotic. Hydrazone of 3-formylrifamycin SV. Selective inhibitor of bacterial DNA-dependent RNA polymerase RNAP.
Catalog No. 50-256-2595
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.