Learn More
AdipoGen Rifamycin AMP-DA
SDP
Supplier:  AdipoGen AGCN20331M010
Encompass_Preferred
Chemical. CAS 16783-99-6. Formula C41H56N4O11. MW 780.9. Semisynthetic. Ansamycin antibiotic. Selective inhibitor of bacterial DNA-dependent RNA polymerase RNAP. Effective against mycobacteria, and are therefore used in research of tuberculosis, leprosy and mycobacterium avium complex MAC infections.
Catalog No. 50-256-2581
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.