Learn More
Honeywell™ Rig Dog™ Thermal-Lined Cold Protect Impact Gloves - Slip On
Catalog No. 19085646 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-085-646 Large
19-085-647 X-Large
19-085-648 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-085-646 Supplier Honeywell™ Supplier No. 43612BY/9L
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Gloves provide impact protection and comfort, while also enhancing grip on oily surfaces. The EN 511 winter lining protects hands from temperatures down to -20°F.
  • TPR padding on the back of the glove provides ANSI/ISEA level 2 impact protection
  • Honeycomb structure enhances energy absorption on the back of the hand
  • Polyurethane tread on the palm is ideal for greasy or muddy situations
  • Thermal lining protects hands in temperatures down to -20°F
  • Slip on cuff for easy donning and doffing
Specifications
ANSI Abrasion Rating ANSI 4
Certifications/Compliance ANSI/ISEA 138 Level 2 Impact Protection
Coating Material Uncoated
Color Black, Red, Hi-vis Yellow
Cut Resistance Rating ANSI A6
Hand Specific
Material Polyester, Spandex
Size Large
Glove Cuff Color Black
Glove Cuff Style Open
Series 43-612BY
Temperature (Metric) Min. -20°F