Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Rig Dog™ Mud Grip Gloves

Catalog No. 19085654 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-085-654 X-Large
19-085-652 Medium
19-085-653 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-085-654 Supplier Honeywell™ Supplier No. 42623BO/10XL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Provide the protection of Xtreme with an additional PU tread palm that squeegees thick oily mud from surfaces for a more secure grip.

  • Highly breathable, designed with an emphasis on wearer comfort
  • PU tread palm squeegees thick oily mud from surfaces for a more secure grip
  • Inner layer offers moisture absorption and a quick dry function to quickly absorb sweat and keep hands dry
  • Enhanced, level 2 impact protection from ANSI/ISEA 138
  • Additional layers provide A6/F level cut resistance, and guard against abrasions
  • Ergonomic design and flex-grooves for increased dexterity

Specifications

Certifications/Compliance ANSI/ISEA 138 Level 2
Coating Material Polyurethane
Color Black, Orange
Cut Resistance Rating ANSI A6
Hand Specific
Material TPR
Product Type Mechanic's Gloves
Size X-Large
Finger Style Full
For Use With (Application) Drilling, material handling, rigging, ship building
Model 42-623BO/10XL
Packaging Type Individual Polybag
Series Rig Dog™ Mud Grip
Thumb Style Reinforced
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.