Learn More
Eisco™ Right Angle Refraction Prism
SCI_ED
Catalog No. S27717 Shop All Eisco Products
Change view
Click to view available options
Right Angle
Catalog No.
S27717 Right Angle
1 options
Catalog No. S27717 Supplier Eisco™ Supplier No. PH0573C
Add to Cart
Add to Cart
Acrylic Right-Angle Refraction Prisms are an excellent tool for optical experiments including refraction. Create a nice rainbow by using the simple led light of a smart phone. Just create a small slit in a black piece of paper and use that slit in front of the led which will create a light ray.
  • Right Angle Refraction Prism
  • Made of high quality acrylic
  • Will create a rainbow with any standard light source, ex. LED from a cellphone
  • Designed for physics and educational uses
  • Dimensions: 0.4 x 3.25 x 3.25 in.
Specifications
Product Type Prism, Right-angled Refraction
Material Acrylic
For Use With (Application) Optics, Light Refraction, and Wavelength Experiments
Height (English) 3.25 in.