Learn More
United Scientific Supplies Right Angled Acrylic Prism Set
SDPSCI_ED
Catalog No. S82873 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Material:
Acrylic
Catalog No. Material
S82873 Acrylic
1 options
This item is not returnable. View return policy
Catalog No. S82873 Supplier United Scientific Supplies Supplier No. PAR123
Add to Cart
Add to Cart
This item is not returnable. View return policy
Right Angled Acrylic Prism Set includes a wide assortment for classroom use.
  • Constructed of clear acrylic with highly polished surfaces, these prisms are ideal for projecting a clear, sharp spectrum.
  • Set of Right-Angled prisms includes 25 mm, 50 mm, and 100 mm long prisms.
Specifications
Product Type Prisms
Material Acrylic
Color Clear