Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Right-to-Know Safety-Vented Wash Bottles, GHS Labeling
SCI_ED

Catalog No. S05125 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Change view
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Chemical Label:
Acetone
Assorted
Deionized Water
Distilled Water
Ethanol
Isopropanol
Methanol
Methyl Ethyl Ketone
Sodium Hypochlorite (Bleach)
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Chemical Label
S05125 500 mL Sodium Hypochlorite (Bleach)
S42981 500 mL Methyl Ethyl Ketone
S05123 500 mL Assorted
S05130 500 mL Ethanol
S05129 500 mL Deionized Water
S05120 500 mL Distilled Water
S05122 500 mL Acetone
S05128 500 mL Methanol
S05127 500 mL Isopropanol
S05121 1000 mL Isopropanol
S05124 1000 mL Acetone
S05126 1000 mL Distilled Water
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Catalog No. S05125 Supplier Bel-Art™ Supplier No. F124160015/EMD
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specially designed valve prevents pressure build-up, which eliminates potentially dangerous chemical drips. Allows dispensing of wash bottle in upright or inverted position.

  • Wide mouth 53 mm (2 in.) for easy refilling
  • Translucent LDPE bottles with color-coded caps for easy identification
  • Pre-printed with chemical name and formula, GHS pictogram and signal word, National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment
  • Bottles with labels that do not indicate a hazardous chemical do not have/require GHS pictogram or signal word
  • Methyl Ethyl Ketone and Toluene do not include vented valve
  • Assortment is 4 per bag, includes one each: Acetone, Isopropanol, Bleach, and Ethanol
  • Sodium Hypochlorite is white LDPE for light-protection between 230 and 450 nm
  • Bottles with labels that do not indicate a hazardous chemical do not have/require GHS pictogram or signal word.

Compliance

Globally Harmonized System of Classification and Labeling of Chemicals (GHS) and OSHA's HazCom 2012 requirements–as pertains to Workplace Labels

Specifications

Capacity 500 mL, 16 oz.
Capacity (Metric) 500 mL
Capacity (English) 16 oz.
Color Yellow
Material LDPE
Quantity 4/Pk.
Sterility Non-sterile
Closure Included Yes
Disposable Reusable
Graduated No
Mouth Wide, Threaded
Shape Cylindrical
Product Type Safety-vented Wash Bottle
Chemical Label Sodium Hypochlorite (Bleach)
Autoclavable Not Autoclavable
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.