Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RIM4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563911
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563911 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563911 Supplier Invitrogen™ Supplier No. PA563911
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TSSMVDPATG PLLRQASQLS LESTVGPCGE Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

Regulates synaptic membrane exocytosis. Also a member of a family of synaptic proteins involved in neurotransmitter release.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIM4
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rims4
Gene Accession No. Q9H426
Gene Alias 6530406L02; C20orf190; rab3-interacting molecule 4; regulating synaptic membrane exocytosis 4; regulating synaptic membrane exocytosis protein 4; RIM 4; RIM4; RIM-4; RIM4 gamma; RIM4gamma; RIM4-gamma; RIMS4; synaptic regulatory protein RIM4gamma
Gene Symbols Rims4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RIM4. Recombinant protein control fragment (Product #RP-101981).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140730
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.