Learn More
PIP™ Rimless Safety Glasses
Catalog No. 19153411 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Frame Material:
Polycarbonate
Polycarbonate, PVC
Polycarbonate, PVC, TPR
Lens Coating:
Anti-fog, Scratch-resistant
Anti-reflective, Scratch-resistant
Scratch-resistant
Uncoated
Lens Tint:
Clear
Gray
Frame Color:
Black
Black, Gray
Black, Green
Black, Maroon
Clear
Clear, Blue
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Material Lens Coating Lens Tint Frame Color
19-153-411 Polycarbonate, PVC, TPR Anti-reflective, Scratch-resistant Gray Black, Gray
19-153-413 Polycarbonate Anti-reflective, Scratch-resistant Gray Black, Gray
19-148-254 Polycarbonate Anti-fog, Scratch-resistant Gray Black
19-148-251 Polycarbonate Scratch-resistant Clear Black
19-148-252 Polycarbonate Scratch-resistant Gray Black
19-035-250 Polycarbonate Uncoated Clear Clear
19-148-006 Polycarbonate, PVC Anti-fog, Scratch-resistant Clear Black, Green
19-148-007 Polycarbonate, PVC Anti-reflective, Scratch-resistant Clear Black, Green
19-148-004 Polycarbonate, PVC Anti-fog, Scratch-resistant Clear Black, Maroon
19-148-005 Polycarbonate, PVC Anti-fog, Scratch-resistant Gray Black, Maroon
19-148-253 Polycarbonate Anti-fog, Scratch-resistant Clear Black
19-153-410 Polycarbonate, PVC, TPR Anti-reflective, Scratch-resistant Clear Clear, Blue
19-153-412 Polycarbonate Anti-reflective, Scratch-resistant Clear Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 missing translation for 'options'
This item is not returnable. View return policy
Catalog No. 19-153-411 missing translation for 'mfr' PIP™ missing translation for 'supplierNo' 250320521
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Excellent field of vision with trendy rimless design
  • Lightweight, rimless design provides all day user comfort
  • Thin, flexible temples provide a secure fit without uncomfortable pressure points
  • Molded nose bridge design to fit the vast majority of users
  • 9.5 Base curve lens
  • Meets ANSI Z87.1+
Specifications
Lens Coating Anti-reflective, Scratch-resistant
Description Rubber nosepads
Lens Tint Gray
Product Type Safety Glasses
Frame Color Black, Gray
Temple Style Flexible bayonet temples
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate, PVC, TPR
UV Protection UV400
Frame Type Rimless
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Warning - Prop 65 - Cancer and Reproductive Harm