Learn More
Cayman Chemical Rimonabant, 168273-06-1, 10 mg
Supplier: Cayman Chemical 900048410
Molecular Formula C22H21Cl3N4O, Purity ≥98%, Formula Weight 463.8, Synonyms: SR141716; A potent and selective CB1 receptor agonist (Kis = 5.6 and >1,000 nM for CB1 and CB2, respectively); induces lipolysis, reduces hepatomegaly, decreases body weight, and improves dyslipidemia by reducing triglyceride, free fatty acid, and total cholesterol levels and by increasing HDL/LDL ratios in rodent models; elicits antiproliferative and immunomodulatory effects in vitro.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.