Learn More
Medchemexpress LLC Rimonabant (SR141716) | 168273-06-1 | MFCD04034714 | >98.0% | C22H21Cl3N4O | 10MG
Supplier: Medchemexpress LLC HY1413610MG
Rimonabant (SR141716) is a selective central cannabinoid receptor (CB1) antagonist used as a research standard in pharmacology. It is brain-penetrant, reported to have a Ki of approximately 1.8 nM at CB1, and is supplied as a crystalline solid for in vitro and in vivo studies.
- Selective CB1 receptor antagonist with high affinity (Ki ≈ 1.8 nM).
- Brain penetrant, suitable for central nervous system studies.
- High purity appropriate for research applications (>98%).
- Soluble in DMSO and common organic solvents for assay preparation.
- Used in pharmacology and metabolic research to probe cannabinoid signaling.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.