Learn More
TARGETMOL CHEMICALS INC RIMONABANT 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1519L
Encompass_Preferred
Also available in 10 mg 25 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Rimonabant (SR141716) is an inverse agonist for the cannabinoid receptor CB1. It is an anorectic anti-obesity drug produced and marketed by Sanofi-Aventis. Its main avenue of effect is a reduction in appetite. Rimonabant is the first selective CB1 receptor blocker to be approved for use anywhere in the world. Rimonabant is approved in 38 countries including the E.U. Mexico and Brazil. It was rejected for approval for use in the United States. purity: 99%
Catalog No. 50-257-7360
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.