Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIMS3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501729968 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-9968 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-9968 Supplier Proteintech Group Inc Supplier No. 150881AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Rab3-interacting molecules (RIMs) are synaptic proteins are synaptic proteins necessary for neural transmission and plasticity. While both Rim1 and Rim 2 are thought to be effector proteins for Rab3, binding to Rab3 on synaptic vesicles in a GTP-dependent manner, less is known of Rim3. Expression of Rim3 in PC12 cells induced a significant increase in calcium-triggered exocytosis, with no appreciable change in the baseline release, suggesting that it plays a role in the regulation of exocytosis. Rim3 protein localizes primarily to neuronal dendrites and the postsynaptic densities, as opposed to Rim1 which is found in presynapse locations, indicating that Rim3 may contribute to synapse transmission and plasticity.

Specifications

Antigen RIMS3
Applications Western Blot
Classification Polyclonal
Concentration 0.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RIMS3
Gene Accession No. Q80U57, Q9JIR3, Q9UJD0
Gene Alias KIAA0237, NIM3, Rab 3 interacting molecule 3, RIM 3, RIM3, RIM3 gamma, RIMS3
Gene Symbols Rims3
Host Species Rabbit
Immunogen RIMS3 Fusion Protein Ag7171
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 242662, 65025, 9783
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.