Learn More
Ansell RINGERS™ R065 Cut-Resistant Gloves
Catalog No. 17103090 Shop All Ansell Products
Change view
Click to view available options
Size:
8
9
10
11
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-103-090 8
17-103-091 9
17-103-092 10
17-103-093 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17-103-090 Supplier Ansell Supplier No. 065G08
Only null left
Add to Cart
Add to Cart
Request a Quote
RINGERS™ R065 Cut-resistant impact gloves, with a TPR design and knit shell for dexterous comfort.
  • Breathable knit shell: Cut resistance, dexterity and comfort
  • TPR material: For protective gloves that ensure impact protection
  • Touchscreen compatibility: Enhanced safety and usability
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Ecovadis label, EN 388:2016 +A1:2018, EN407:2020, EN ISO 21420:2020, ANSI Cut A4, ANSI ABR 4, ANSI/ISEA 105-2016 PUN 4, ANSI/ISEA 138-2, Category III
Coated Area Full Dipped
Construction Knit
Cuff Material HPPE
Description Specialized defenses: Thanks to a breathable knit shell developed with RINGERS™ F3 Technology™, these impact-resistant gloves combine advanced, EN/ANSI-certified cut resistance with excellent comfort and dexterity.
Coating Material Nitrile
Color Gray
Cut Resistance Rating ANSI A4
Hand Specific
Latex Free Yes
Material HPPE
Product Type Mechanical Protection Gloves
Size 8
EN388 Standard EN388:4X43DP
For Use With (Application) Makeup and breakout tools, Running tools into the wellbore, Engagement and release of drill string slips, Preparation of site - set-up and disassembly of tool string, General pumping work, Operating of heavy machinery, Equipment maintenance and repair, Pipe handling, Handling parts with sharp, rough edges, Scaffolding, Loading and hauling, Assembling and mounting components
Gender Unisex
Glove Cuff Color Gray
Glove Cuff Style Knit
Grip Design Sandy Nitrile
Liner Material HPPE
Lining Knit Lining
Model R065
Packaging Type 12 Pairs/Bag, 6 Bags/Carton
Thumb Style Reinforced
Yarn Gauge 13